missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,3-dimethylacrylate, 98%

Catalog No. AAL1973118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1973118 50 g
AAL1973109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1973118 Supplier Thermo Scientific Chemicals Supplier No. L1973118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 924-50-5 | C6H10O2 | 114.144 g/mol

Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,3-dimethylacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in chloroform. Insoluble in water,

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is sensitive to light.

Chemical Identifiers

CAS 924-50-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00043940
InChI Key FZIBCCGGICGWBP-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-butenoate, methyl 3,3-dimethylacrylate, 2-butenoic acid, 3-methyl-, methyl ester, 3-methyl-but-2-enoic acid methyl ester, methyl senecioate, 3,3-dimethylacrylic acid methyl ester, methyl-3,3-dimethylacrylate, crotonic acid, 3-methyl-, methyl ester, methyl-3,3-dimethyl acrylate, methylsenecioate
PubChem CID 13546
IUPAC Name methyl 3-methylbut-2-enoate
SMILES CC(=CC(=O)OC)C

Specifications

Melting Point -41°C
Density 0.936
Boiling Point 134°C to 139°C
Flash Point 33°C (91°F)
Refractive Index 1.4375
Linear Formula (CH3)2C=CHCO2CH3
Quantity 50 g
UN Number UN3272
Beilstein 1741592
Sensitivity Light sensitive
Solubility Information Soluble in chloroform. Insoluble in water,
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material Methyl 3,3-dimethylacrylate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 213-107-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only