missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,3-Dimethylacrylate 98.0+%, TCI America™

Catalog No. M205225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M205225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M205225G Supplier TCI America Supplier No. M205225G

Chemical Identifiers

CAS 924-50-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00043940
InChI Key FZIBCCGGICGWBP-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-butenoate, methyl 3,3-dimethylacrylate, 2-butenoic acid, 3-methyl-, methyl ester, 3-methyl-but-2-enoic acid methyl ester, methyl senecioate, 3,3-dimethylacrylic acid methyl ester, methyl-3,3-dimethylacrylate, crotonic acid, 3-methyl-, methyl ester, methyl-3,3-dimethyl acrylate, methylsenecioate
PubChem CID 13546
IUPAC Name methyl 3-methylbut-2-enoate
SMILES CC(=CC(=O)OC)C

Specifications

Color Colorless
Boiling Point 136°C
Quantity 25 g
UN Number 3272
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3,3-Dimethylacrylate
EINECSNumber (2)-3496
TSCA Yes