accessibility menu, dialog, popup

UserName

CAS RN 921-53-9

KKOH(R)(R)OHOOOO

CAS RN 921-53-9

IUPAC Name: dipotassium (2R,3R)-2,3-dihydroxybutanedioate
Synonyms: potassium tartrate|argol|dipotassium tartrate|potassium L-tartrate
Molecular Weight (g/mol): 226.27
Molecular Formula: C4H4K2O6
InChi Key: AVTYONGGKAJVTE-OLXYHTOASA-L
SMILES: [K+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)