accessibility menu, dialog, popup

UserName

CAS RN 91983-14-1

IUPAC Name: [2-(bromomethyl)phenyl]boronic acid
Synonyms: 2-(bromomethyl)phenylboronic acid
Molecular Weight (g/mol): 214.85
Molecular Formula: C7H8BBrO2
InChi Key: MYVJCOQGXCONPE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1CBr
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results