missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B36001G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36001G Supplier TCI America Supplier No. B36001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91983-14-1
Molecular Formula C7H8BBrO2
Molecular Weight (g/mol) 214.85
MDL Number MFCD01318941
InChI Key MYVJCOQGXCONPE-UHFFFAOYSA-N
Synonym 2-bromomethylphenylboronic acid, 2-bromomethyl benzeneboronic acid, 2-bromomethyl phenylboronic acid, 2-bromomethyl phenyl boronic acid, 2-bromomethylphenylboronicacid, 2-boronobenzyl bromide, o-boronobenzyl bromide, 2-bromomethylphenyl boronic acid, pubchem7781
PubChem CID 2773278
IUPAC Name [2-(bromomethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1CBr

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 214.85
Physical Form Crystalline Powder
Chemical Name or Material 2-(Bromomethyl)phenylboronic Acid (contains varying amounts of Anhydride)
TSCA No
Recommended Storage Refrigerator