missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)phenylboronic acid, 95%, Thermo Scientific Chemicals

Catalog No. AC445330050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445330050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445330050 Supplier Thermo Scientific Chemicals Supplier No. 445330050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91983-14-1 | Molecular Formula: C7H8BBrO2 | Molecular Weight: 214.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91983-14-1
Molecular Formula C7H8BBrO2
Molecular Weight (g/mol) 214.85
MDL Number MFCD01318941
InChI Key MYVJCOQGXCONPE-UHFFFAOYSA-N
Synonym 2-bromomethylphenylboronic acid, 2-bromomethyl benzeneboronic acid, 2-bromomethyl phenylboronic acid, 2-bromomethyl phenyl boronic acid, 2-bromomethylphenylboronicacid, 2-boronobenzyl bromide, 2-bromomethylphenyl boronic acid, o-boronobenzyl bromide, pubchem7781
PubChem CID 2773278
IUPAC Name [2-(bromomethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1CBr

Specifications

Melting Point 135°C to 136.5°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 214.85
Percent Purity 95%
Chemical Name or Material 2-(Bromomethyl)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only