accessibility menu, dialog, popup

UserName

(S)(R)(S)O(R)(R)OHHOOO(S)(R)(R)O(R)(S)∗OHHOHO∗O∗mn

CAS RN 9004-54-0

IUPAC Name: Dextran
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C6H9O5)n(C6H10O5)m
InChi Key: —
SMILES: O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
Quantity 
  • (9)
  • (9)
  • (1)
  • (18)
  • (7)
  • (2)
  • (10)
  • (1)
  • (12)
Grade 
  • (12)
  • (3)
  • (5)
  • (1)
  • (19)
Technique 
  • (12)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (23)
  • (20)
  • (1)
  • (13)
  • (12)
  • (20)
  • (4)
  • (32)
  • (9)
  • (9)
  • (1)
  • (18)
  • (7)
  • (2)
  • (10)
  • (1)
  • (12)
  • (12)
  • (3)
  • (5)
  • (1)
  • (19)
  • (12)