missing translation for 'onlineSavingsMsg'
Learn More

Dextran, MW ca 40,000

Catalog No. AAJ6369036
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6369036 500 g
AAJ6369009 10 g
AAJ6369018 50 g
AAJ6369022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6369036 Supplier Thermo Scientific Chemicals Supplier No. J6369036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dextran, CAS # 9004-54-0, is a glucose polymer. It is mainly used to increase electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity). | CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol

Dextran is used in a bead form to aid in bioreactor applications, some size-exclusion chromatography matrices and in osmotic stress technique study involved in biological molecules. It can be used as a stabilizing coating to protect metal nanoparticles from oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Dextran is a glucose polymer biosynthesized by the non-pathogenic Leuconostoc mesenteroides

Application

  • Dextran is a complex branched glucan that can be used to enhance electronegativity and reduce erythrocyte aggregation and platelet adhesiveness (anticoagulant activity)
  • This compound promotes the activation of plasminogen by inhibiting α-2 antiplasmin
  • Dextran can be used to reduce thrombosis in animal models
  • It is used in osmotic stress techniques where osmotic pressure is applied to biological molecules
  • Dextran can be used as an immobilization agent in biosensors

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
Molecular Weight (g/mol) 504.44
MDL Number MFCD00130935
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
PubChem CID 4125253
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Merck Index 14,2948
Solubility Information Soluble in water,dimethyl sulfoxide,ethylene glycol and glycerol.
Formula Weight ≈40 kd
Chemical Name or Material Dextran
EINECSNumber 232-677-5
RTECSNumber HH9246000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only