missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, MW ca 250,000
Description
Dextran, is used as a multi-use, branched glucan. Low molecular weight dextran is useful as a stabilizer, volume extender, lubricant, binding platform, and matrix component. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Merck Index | 14,2948 |
| Solubility Information | It is soluble in water (100mg/ml),DMSO (freely),formamide (freely),ethylene glycol (freely),and glycerol (freely). |
| Formula Weight | ≈250 kd |
| Chemical Name or Material | Dextran |
Safety and Handling
| EINECSNumber | 232-677-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only