accessibility menu, dialog, popup

UserName

∗∗OHn

CAS RN 9002-89-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C2H4O)n
InChi Key: —
SMILES: OC(-*)C-*
Molecular Weight (g/mol) 
  • (1)
  • (64)
  • (1)
  • (1)
  • (1)
Quantity 
  • (17)
  • (13)
  • (1)
  • (5)
  • (13)
  • (2)
  • (3)
  • (1)
  • (1)
  • (11)
  • (1)
Percent Purity 
  • (6)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (9)
  • (1)
  • (2)
  • (3)
Grade 
  • (4)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (30)
  • (1)
  • (3)
  • (3)
  • (10)
  • (3)
  • (48)
  • (1)
  • (64)
  • (1)
  • (1)
  • (1)
  • (17)
  • (13)
  • (1)
  • (5)
  • (13)
  • (2)
  • (3)
  • (1)
  • (1)
  • (11)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (9)
  • (1)
  • (2)
  • (3)
  • (4)
  • (6)
  • (1)