missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 99-100% hydrolyzed, approx. M.W. 124,000, Thermo Scientific™

Catalog No. AC458230050
Change view
Click to view available options
Quantity:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458230050 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458230050 Supplier Thermo Scientific Chemicals Supplier No. 458230050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 9002-89-5
Quantity 5kg
Molecular Formula C2H4O
Linear Formula [-CH2CH(OH)-]n
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
Solubility Information Solubility in water: soluble in hot water.
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
SMILES C=CO
IUPAC Name ethenol
Molecular Weight (g/mol) 44.053
PubChem CID 11199
Chemical Name or Material Poly(vinyl alcohol)
Show More Show Less

RUO – Research Use Only