missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Color | White to Yellow |
| pH | 4.5 to 6.5 (4% aq. soln.) |
| Physical Form | Powder |
| Melting Point | 160.0°C to 240.0°C |
| Ash | 90%% (Na2O) max. |
| Quantity | 500 g |
| Infrared Spectrum | Authentic |
| Packaging | Plastic bottle |
| Linear Formula | [-CH2CH(OH)-]n |
| Viscosity | 3.5 to 4.5cP (4% aq. solution, 20°C) Brookfield |
| Show More |
RUO – Research Use Only