accessibility menu, dialog, popup

UserName

∗∗n

CAS RN 9002-88-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C2H4)n
InChi Key: —
SMILES: *-CC-*
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (17)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)