missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene for GPC 2,000, MilliporeSigma™ Supelco™

Catalog No. 111015772
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015772 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015772 Supplier Merck Emd Millipore Supplier No. 81223100MG

Specifications

CAS 9002-88-4
Physical Form Neat
Linear Formula H(CH2 CH2)nH
MDL Number MFCD00084423
Type Analytical Standard
Quantity 100 mg
Grade GPC