accessibility menu, dialog, popup

UserName

H3COOH3C

CAS RN 87-24-1

IUPAC Name: ethyl 2-methylbenzoate
Synonyms: ethyl 2-methylbenzoate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: SOUAXOGPALPTTC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)