missing translation for 'onlineSavingsMsg'
Learn More

Ethyl o-toluate, 98+%

Catalog No. AAL0059822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0059822 100 g
AAL0059814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0059822 Supplier Thermo Scientific Chemicals Supplier No. L0059822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 87-24-1 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-24-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009112
InChI Key SOUAXOGPALPTTC-UHFFFAOYSA-N
Synonym ethyl o-toluate, ethyl o-methylbenzoate, ethyl orthotoluate, 2-methylbenzoic acid ethyl ester, benzoic acid, 2-methyl-, ethyl ester, ethyl-o-toluate, 2-methyl-benzoic acid ethyl ester, o-toluic acid ethyl ester, ethyl-2-methylbenzoate, ethylo-toluate
PubChem CID 66598
IUPAC Name ethyl 2-methylbenzoate
SMILES CCOC(=O)C1=CC=CC=C1C

Specifications

Density 1.033
Boiling Point 227°C
Flash Point 91°C (195°F)
Refractive Index 1.507
Quantity 100 g
Beilstein 2042517
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material Ethyl o-toluate
Hazard Category H227-H318
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P305+P351+P338-P310-P370+P378q-P501c
EINECSNumber 201-734-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only