missing translation for 'onlineSavingsMsg'
Learn More

Ethyl o-Toluate 98.0+%, TCI America™

Catalog No. T0294500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0294500G 500 g
T029425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0294500G Supplier TCI America Supplier No. T0294500G

Chemical Identifiers

CAS 87-24-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009112
InChI Key SOUAXOGPALPTTC-UHFFFAOYSA-N
Synonym ethyl o-toluate, ethyl o-methylbenzoate, ethyl orthotoluate, 2-methylbenzoic acid ethyl ester, benzoic acid, 2-methyl-, ethyl ester, ethyl-o-toluate, 2-methyl-benzoic acid ethyl ester, o-toluic acid ethyl ester, ethyl-2-methylbenzoate, ethylo-toluate
PubChem CID 66598
IUPAC Name ethyl 2-methylbenzoate
SMILES CCOC(=O)C1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 227°C
Quantity 500 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl o-Toluate
EINECSNumber (3)-1380
TSCA Yes