accessibility menu, dialog, popup

UserName

FFBr

CAS RN 85117-99-3

IUPAC Name: 2-(bromomethyl)-1,4-difluorobenzene
Synonyms: 2-(bromomethyl)-1,4-difluorobenzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: ONWGSWNHQZYCFK-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(CBr)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)