missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. D25975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25975G 5 g
D259725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25975G Supplier TCI America Supplier No. D25975G
Only null left

Chemical Identifiers

CAS 85117-99-3
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD00009897
InChI Key ONWGSWNHQZYCFK-UHFFFAOYSA-N
Synonym 2,5-difluorobenzyl bromide, 2-bromomethyl-1,4-difluorobenzene, 2,5-difluorobenzylbromide, alpha-bromo-2,5-difluorotoluene, benzene, 2-bromomethyl-1,4-difluoro, timtec-bb sbb006680, 2-bromomethyl-1,4-difluoro-benzene, pubchem10213, 2,2-difluorobenzylbromide, acmc-209q4i
PubChem CID 522830
IUPAC Name 2-(bromomethyl)-1,4-difluorobenzene
SMILES FC1=CC=C(F)C(CBr)=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 2924
Formula Weight 207.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluorobenzyl Bromide