missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Difluorobenzyl bromide, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.609 |
| Refractive Index | 1.526 |
| Quantity | 50 g |
| UN Number | UN3265 |
| Beilstein | 7089248 |
| Formula Weight | 207.02 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,5-Difluorobenzyl bromide |
Safety and Handling
| Hazard Category | H225-H302-H314 |
| Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: II |
| EINECSNumber | 285-651-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only