accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 84-75-3

IUPAC Name: 1,2-dihexyl benzene-1,2-dicarboxylate
Synonyms: di-n-hexyl phthalate hexyl phthalate, 1,2- 1,2-hexyl phthalate N-dihexyl phthalate dihexylester kyseliny ftalove di-n-hexylphthalate
Molecular Weight (g/mol): 334.46
Molecular Formula: C20H30O4
InChi Key: KCXZNSGUUQJJTR-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)