missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals

Catalog No. AAA1259330
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1259330 250 g
AAA1259309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1259330 Supplier Thermo Scientific Chemicals Supplier No. A1259330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalates are by far the most widely used plasticizers, primarily to make soft and flexible polyvinyl chloride (PVC) for the applications in the industry of automotive, building & construction material, cable, flooring, medical device and toys. Minor quantity of phthalates are used in adhesives, caulk, sealants, paint to improve work performance. Small molecule phthalates are used as solvents in perfumes to provide longer linger longer and in nail polish to prevent chipping.

Solubility
Soluble in water (0.24 mg/L).

Notes
Store at 4°C. Avoid excessive heat and light. Reactive towards oxidizing agents.

Chemical Identifiers

CAS 84-75-3
Molecular Formula C20H30O4
Molecular Weight (g/mol) 334.46
MDL Number MFCD00043720
InChI Key KCXZNSGUUQJJTR-UHFFFAOYSA-N
Synonym dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate
PubChem CID 6786
ChEBI CHEBI:34678
IUPAC Name dihexyl benzene-1,2-dicarboxylate
SMILES CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC

Specifications

Density 1.01
Boiling Point 185°C to 187°C (0.5 mmHg)
Refractive Index 1.487
Quantity 250 g
UN Number UN3082
Solubility Information Soluble in water (0.24mg/L).
Formula Weight 334.46
Percent Purity 97%
Chemical Name or Material Di-n-hexyl phthalate
Hazard Category H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 201-559-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only