missing translation for 'onlineSavingsMsg'
Learn More

Dihexyl Phthalate 98.0+%, TCI America™

Catalog No. P13445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13445G 5 g
P134425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13445G Supplier TCI America Supplier No. P13445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84-75-3
Molecular Formula C20H30O4
Molecular Weight (g/mol) 334.46
MDL Number MFCD00043720
InChI Key KCXZNSGUUQJJTR-UHFFFAOYSA-N
Synonym dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate
PubChem CID 6786
ChEBI CHEBI:34678
IUPAC Name 1,2-dihexyl benzene-1,2-dicarboxylate
SMILES CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC

Specifications

Color Yellow
Boiling Point 333°C
Quantity 5 g
Formula Weight 334.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dihexyl Phthalate
EINECSNumber (3)-1307
RTECSNumber TI1100000
TSCA Yes