accessibility menu, dialog, popup

UserName

H3COOOHOOOHOHOHOHOHHO

CAS RN 82494-09-5

IUPAC Name: 2-{[6-(decyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-{[6-(decyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 482.57
Molecular Formula: C22H42O11
InChi Key: WOQQAWHSKSSAGF-UHFFFAOYNA-N
SMILES: CCCCCCCCCCOC1OC(CO)C(OC2OC(CO)C(O)C(O)C2O)C(O)C1O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results