missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ n-Decyl-β-D-maltopyranoside, ULTROL™ Grade, Calbiochem™,

Catalog No. 2527185GM
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2527185GM 5 g Glass Bottle
2527181GM 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2527185GM Supplier MilliporeSigma™ Supplier No. 2527185GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82494-09-5
Molecular Formula C22H42O11
Molecular Weight (g/mol) 482.567
InChI Key WOQQAWHSKSSAGF-WXFJLFHKSA-N
PubChem CID 5288728
ChEBI CHEBI:67097
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-decoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥99% (TLC)