missing translation for 'onlineSavingsMsg'
Learn More

Decyl beta-D-maltopyranoside, 97+%

Catalog No. AAJ66644ME
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66644ME 500 mg
AAJ6664403 1 g
AAJ6664406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66644ME Supplier Thermo Scientific Chemicals Supplier No. J66644ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82494-09-5 | C22H42O11 | 482.567 g/mol

Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decyl beta-D-maltopyranoside is used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography. It aids in solubilizing the active cytochrome oxidase from mitochondria.

Solubility
Soluble in methanol at 50mg/ml

Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 82494-09-5
Molecular Formula C22H42O11
Molecular Weight (g/mol) 482.567
MDL Number MFCD00061624
InChI Key WOQQAWHSKSSAGF-WXFJLFHKSA-N
Synonym decyl beta-d-maltopyranoside, decyl 4-o-a-d-glucopyranosyl-b-d-glucopyranoside, n-decyl-b-d-maltoside, decyl-, a-d-maltoside, decyl-beta-d-maltopyranoside, n-decyl beta-d-maltopyranoside, decyl 4-o-alpha-d-glucopyranosyl-beta-d-glucopyranoside, decyl alpha-d-glucopyranosyl-1->4-beta-d-glucopyranoside, n-decyl-beta-d-maltopyranoside, decyl b-d-maltopyranoside
PubChem CID 5288728
ChEBI CHEBI:67097
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-decoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Color White
Quantity 500 mg
Beilstein 4923334
Solubility Information Soluble in methanol at 50mg/ml
Formula Weight 482.56
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Decyl beta-D-maltopyranoside
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only