CAS RN 8042-47-5
Mineral Oil, Light (NF/FCC), Fisher Chemical™
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00131611 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L Synonym: acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Mineral oil, pure
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L Synonym: acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Mineral Oil, Heavy (USP/FCC), Fisher Chemical™
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00131611 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L Synonym: acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Matrix oil, 75 centistoke hydrocarbon oil, Thermo Scientific Chemicals
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00131611 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Matrix oil, 75 centistoke hydrocarbon oil
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00131611 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Mineral oil, high purity
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00131611 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L Synonym: acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Viscosity standard, Specpure™, nominally 120cSt @ 40 and 20cSt @ 100
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00084411 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Viscosity standard, Specpure™, nominally 30cSt at 40° and 5.3cSt at 100°
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00084411 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Viscosity standard, Specpure™, nominally 10cSt @ 40 and 2.7cSt @ 100
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00084411 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 MDL Number: MFCD00131611 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L Synonym: acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Mineral oil, MP Biomedicals™
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L Synonym: acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Kaydol™/Hexadecane, 65/35 v/v, Reagents
CAS: 8042-47-5
Mineral Oil MilliporeSigma™
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L Synonym: acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Mineral Oil, Molecular Biology Grade, Hoefer™
CAS: 8042-47-5 Molecular Formula: C16H10N2Na2O7S2 Molecular Weight (g/mol): 452.363 InChI Key: AEOVEGJBKQQFOP-DDVLFWKVSA-L Synonym: acid orange 10,wool orange 2g,orange g,c.i. acid orange 10,c.i. orange g,c.i. food orange 4,light orange g,colacid orange g,dolkwal orange g,hexacol orange g PubChem CID: 9566064 IUPAC Name: disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate SMILES: C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
HYDRANAL™-Water Standard Oil, Solstice
Oil-based standard specifically designed for accurate Karl Fischer titration with a water content of less than 50 ppm.
- 1
- 2