missing translation for 'onlineSavingsMsg'
Learn More

Viscosity standard, Specpure™, nominally 10cSt @ 40 and 2.7cSt @ 100

Catalog No. AA47036AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47036AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA47036AP Supplier Thermo Scientific Chemicals Supplier No. 047036AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00084411
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Odor Odorless
Flash Point >110°C (230°F)
Physical Form Liquid
Quantity 500 mL
Grade Specpure
Chemical Name or Material Viscosity standard, Nominally 10cSt at 40 and 2.7cSt at 100
Hazard Category H304
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 232-455-8
RTECSNumber PY8047000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only