missing translation for 'onlineSavingsMsg'
Learn More

Kaydol™/Hexadecane, 65/35 v/v, Reagents

Catalog No. 01334893
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334893 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334893 Supplier Labchem Inc Supplier No. CK000300500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kaydol™/Hexadecane, 65/35 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8042-47-5 , 544-76-3

Specifications

Color Colorless
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 0.823
Grade Reagent
Physical Form Liquid
Chemical Name or Material Kaydol™/Hexadecane, 65/35 v/v