accessibility menu, dialog, popup

UserName

CH3SiH3CCH3NS

CAS RN 79265-30-8

IUPAC Name: 2-(trimethylsilyl)-1,3-thiazole
Synonyms: 2-(trimethylsilyl)-1,3-thiazole
Molecular Weight (g/mol): 157.31
Molecular Formula: C6H11NSSi
InChi Key: VJCHUDDPWPQOLH-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=NC=CS1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)