missing translation for 'onlineSavingsMsg'
Learn More

2-Trimethylsilylthiazole 95.0+%, TCI America™

Catalog No. T12995ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12995ML 5 mL
T12991ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12995ML Supplier TCI America Supplier No. T12995ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79265-30-8
Molecular Formula C6H11NSSi
Molecular Weight (g/mol) 157.31
MDL Number MFCD00066274
InChI Key VJCHUDDPWPQOLH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq
PubChem CID 588453
IUPAC Name 2-(trimethylsilyl)-1,3-thiazole
SMILES C[Si](C)(C)C1=NC=CS1

Specifications

Color Yellow
Boiling Point 57°C
Quantity 5 mL
Formula Weight 157.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Trimethylsilylthiazole
TSCA No
Recommended Storage Refrigerator