missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)thiazole, 97%

Catalog No. AAB2190314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2190314 25 g
AAB2190306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2190314 Supplier Thermo Scientific Chemicals Supplier No. B2190314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79265-30-8 | C6H11NSSi | 157.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79265-30-8
Molecular Formula C6H11NSSi
Molecular Weight (g/mol) 157.31
MDL Number MFCD00066274
InChI Key VJCHUDDPWPQOLH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq
PubChem CID 588453
IUPAC Name trimethyl(1,3-thiazol-2-yl)silane
SMILES C[Si](C)(C)C1=NC=CS1

Specifications

Density 0.985
Boiling Point 53°C to 55°C (10 mmHg)
Flash Point 57°C (135°F)
Refractive Index 1.497
Quantity 25 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 157.31
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilyl)thiazole
Hazard Category H227
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only