accessibility menu, dialog, popup

UserName

CAS RN 78755-81-4

IUPAC Name: ethyl 12-fluoro-8-methyl-9-oxo-2,4,8-triazatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaene-5-carboxylate
Synonyms: flumazenil flumazenilo anexate lanexat flumazenil preparation flumazenil (substance) hoffman la roche brand of flumazenil mazicon romazicon flumazenilum
Molecular Weight (g/mol): 303.29
Molecular Formula: C15H14FN3O3
InChi Key: OFBIFZUFASYYRE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2CN(C)C(=O)C3=CC(F)=CC=C3N2C=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)