missing translation for 'onlineSavingsMsg'
Learn More

Flumazenil, 98%

Catalog No. AAJ62537ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62537ME 500 mg
AAJ62537MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62537ME Supplier Thermo Scientific Chemicals Supplier No. J62537ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Benzodiazepine antagonist | CAS: 78755-81-4 | C15H14FN3O3 | 303.293 g/mol

Benzodiazepine antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78755-81-4
Molecular Formula C15H14FN3O3
Molecular Weight (g/mol) 303.293
MDL Number MFCD00242764
InChI Key OFBIFZUFASYYRE-UHFFFAOYSA-N
Synonym flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
PubChem CID 3373
ChEBI CHEBI:5103
IUPAC Name ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
SMILES CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C

Specifications

Color White
Quantity 500 mg
Merck Index 14,4135
Solubility Information Soluble in DMSO to 25mM
Formula Weight 303.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Flumazenil
RTECSNumber NI2922170
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only