missing translation for 'onlineSavingsMsg'
Learn More

Flumazenil 99.0+%, TCI America™

Catalog No. F095825MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F095825MG 25 mg
F0958100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F095825MG Supplier TCI America Supplier No. F095825MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78755-81-4
Molecular Formula C15H14FN3O3
Molecular Weight (g/mol) 303.293
MDL Number MFCD00242764
InChI Key OFBIFZUFASYYRE-UHFFFAOYSA-N
Synonym flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
PubChem CID 3373
ChEBI CHEBI:5103
IUPAC Name ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
SMILES CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C

Specifications

Melting Point 200°C
Color White
Quantity 25 mg
Formula Weight 303.29
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Flumazenil
RTECSNumber N12922170
TSCA No
Recommended Storage Refrigerator