CAS RN 7783-83-7
Ferric Ammonium Sulfate Dodecahydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): 482.176 MDL Number: MFCD00150004 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ammonium iron(III) sulfate dodecahydrate, 98+%
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): 482.176 MDL Number: MFCD00150004 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ammonium iron(III) sulfate dodecahydrate, ACS, 98.5-102.0%
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): 482.176 MDL Number: MFCD00150004 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ammonium iron(III) sulfate dodecahydrate, 99%, ACS reagent
CAS: 7783-83-7 Molecular Formula: H4FeNO8S2·12H2O Molecular Weight (g/mol): 482.19 MDL Number: MFCD00150004 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ferric Ammonium Sulfate, 12-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): 482.176 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis
CAS: 7783-83-7 Molecular Formula: H4FeNO8S2·12H2O Molecular Weight (g/mol): 482.19 MDL Number: MFCD00150004 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ferric Ammonium Sulfate, For Chloride (Volhard's Indicator), Certified, 8%, LabChem™
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): 482.176 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ferric Ammonium Sulfate, 0.0500 N, Ricca Chemical
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): 482.18 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: iron(3+) ammonium dodecahydrate disulfate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Ferric Ammonium Sulfate, 3% (w/v), Ricca Chemical
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): 482.176 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ferric Ammonium Sulfate (Ferric Alum), Saturated, Ricca Chemical
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): 482.176 InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K Synonym: ammonium ferric sulfate,ferric ammonium sulfate dodecahydrate,iron ammonium sulfate dodecahydrate,sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate,ferric ammonium sulfate, acs,azanium iron 3+ disulfate dodecahydrate,ferric iron ammonium dodecahydrate disulfate,azanium iron +3 cation disulfate dodecahydrate,ammonium iron iii sulfate dodecahydrate, acs 100g PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Ferric Ammonium Sulfate, 0.200 N (N/5), Ricca Chemical
Chemical Name or Material: Ferric Ammonium Sulfate CAS: 7783-83-7 CAS Min %: 8.57 CAS Max %: 8.92 CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 92.0 CAS: 7664-93-9 CAS Min %: 0.82 CAS Max %: 0.85
Ferric Ammonium Sulfate, 0.125 N (N/8), Ricca Chemical
Chemical Name or Material: Ferric Ammonium Sulfate CAS: 7783-83-7 CAS Min %: 5.53 CAS Max %: 5.75 CAS: 7732-18-5 CAS Min %: 92.0 CAS Max %: 95.0 CAS: 7664-93-9 CAS Min %: 0.84 CAS Max %: 0.88
Ferric Alum Indicator, Volhard, Ricca Chemical
CAS: 7783-83-7 Molecular Formula: FeH28NO20S2 Molecular Weight (g/mol): Mixture InChI Key: LCPUDZUWZDSKMX-UHFFFAOYSA-K PubChem CID: 197097 IUPAC Name: azanium;iron(3+);disulfate;dodecahydrate SMILES: [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]