missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate Dodecahydrate ACS MP Biomedicals

Catalog No. ICN19404425
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19404425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19404425 Supplier MP Biomedicals Inc Supplier No. 0219404425

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS grade reagent used as a mordant in dyeing, an analytical iron standard, and a catalyst.

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 482.2
Grade ACS
Density 1.71g/mL
Chemical Name or Material Ferric Ammonium Sulfate, Dodecahydrate