missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis

Catalog No. AC205885000
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205885000 500 g Plastic bottle
AC205880500 50 g Plastic bottle
AC205881000 100 g Plastic bottle
AC205880025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC205885000 Supplier Thermo Scientific Chemicals Supplier No. 205885000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7783-83-7 | H4FeNO8S2·12H2O | 482.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-83-7
Molecular Formula H4FeNO8S2·12H2O
Molecular Weight (g/mol) 482.19
MDL Number MFCD00150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Boiling Point 230.0°C
Melting Point 39.0°C to 41.0°C
Color Gray-Purple
Physical Form Adhering Crystalline Powder or Crystals
Quantity 500 g
Assay Percent Range 99% min. (Iodometry)
Linear Formula (NH4)Fe(SO4)2·12H2O
Merck Index 15, 515
Solubility Information Solubility in water: 1240g/L water (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 482.19
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.71
Density 1.7100g/mL
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
RTECSNumber WS5900000

RUO – Research Use Only