missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Ammonium Sulfate, For Chloride (Volhard's Indicator), Certified, 8%, LabChem™
Specifications
Specifications
| CAS Max % | 92, 8 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Suitability | Passes Test |
| Linear Formula | FeNH4S2O8·12H2O |
| Decomposition Information | Sulfur compounds; Gaseous Ammonia; Nitrogen oxides |
| Color | Brown/Yellow |
| Solubility Information | Soluble in water |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |