missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulphate Solution R5, Ricca Chemical

Catalog No. R3058500500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R3058500500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R3058500500 Supplier Ricca Chemical Company Supplier No. R3058500500B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-83-7 , 7697-37-2 , 7732-18-5
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.18
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097  
IUPAC Name iron(3+) ammonium dodecahydrate disulfate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Golden
CAS Min % 15.47, 30.98, 52.0
CAS Max % 16.1, 32.25, 54.0
Packaging Amber Poly
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ferric Ammonium Sulphate Solution, Reagent