missing translation for 'onlineSavingsMsg'
Learn More

Selenium ICP Standard, 10,000 ppm Se in 5% HNO3, Ricca Chemical

Catalog No. p-3252245
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PSE10KN50 Natural Poly Bottle
PSE10KN100 Natural LDPE Bottle
PSE10KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PSE10KN50 Supplier Ricca Chemical Company Supplier No. PSE10KN50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7782-49-2 , 7697-37-2 , 7732-18-5
Molecular Formula Se
Molecular Weight (g/mol) 78.971
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Se
CAS Min % 86.0, 11.57, 0.86
CAS Max % 89.0, 12.05, 0.89
Equivalence 1 mL = 10 mg Se
Chemical Name or Material Selenium ICP Standard