missing translation for 'onlineSavingsMsg'
Learn More

Selenium ICP Standard, 1000 ppm Se in 3% HNO3, Ricca Chemical

Catalog No. PSE1KN500
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PSE1KN500 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSE1KN500 Supplier Ricca Chemical Company Supplier No. PSE1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 7782-49-2, 7697-37-2, 7732-18-5
Concentration or Composition Notes 1,000ppm Se
CAS Min % 93.0, 4.76, 0.09
CAS Max % 97.0, 4.95, 0.1
Equivalence 1 mL = 1 mg Se
Molecular Formula Se
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES [Se]
IUPAC Name selenium
Molecular Weight (g/mol) 78.971
PubChem CID 6326970
ChEBI CHEBI:27568
Chemical Name or Material Selenium ICP Standard
Show More Show Less