missing translation for 'onlineSavingsMsg'
Learn More

Selenium ICP Standard, 1000 ppm Se in 3% HNO3, Ricca Chemical

Catalog No. PSE1KN100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PSE1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSE1KN100 Supplier Ricca Chemical Company Supplier No. PSE1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7782-49-2 , 7697-37-2 , 7732-18-5
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Se
CAS Min % 93.0, 4.76, 0.09
CAS Max % 97.0, 4.95, 0.1
Equivalence 1 mL = 1 mg Se
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Selenium ICP Standard
Show More Show Less