missing translation for 'onlineSavingsMsg'
Learn More

Selenium ingot/button,≈38mm (1.5 in.) dia.

Catalog No. AA45115KF
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45115KF 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45115KF Supplier Thermo Scientific Chemicals Supplier No. 045115KF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7782-49-2 | Se | 78.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-49-2
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]

Specifications

Physical Form Pieces
Quantity 1 Pc.
Odor Odorless
Chemical Name or Material Selenium ingot/button, ≈38mm (1.5 in.) dia.
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 231-957-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only