accessibility menu, dialog, popup

UserName

H3COCH3OOCH3

CAS RN 776-99-8

IUPAC Name: 1-(3,4-dimethoxyphenyl)propan-2-one
Synonyms: 1-(3,4-dimethoxyphenyl)propan-2-one
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: UMYZWICEDUEWIM-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(C)=O)C=C1OC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)