missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetone 97.0+%, TCI America™

Catalog No. V0052500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0052500ML 500 mL
V005225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0052500ML Supplier TCI America Supplier No. V0052500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 776-99-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008772
InChI Key UMYZWICEDUEWIM-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
PubChem CID 69896
IUPAC Name 1-(3,4-dimethoxyphenyl)propan-2-one
SMILES COC1=CC=C(CC(C)=O)C=C1OC

Specifications

Color Yellow
Boiling Point 167°C
Quantity 500 mL
Formula Weight 194.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethoxyphenylacetone
EINECSNumber (8)-0683
RTECSNumber UC1795500
TSCA No