missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetone, 97%

Catalog No. AAL0547718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0547718 50 g
AAL0547709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0547718 Supplier Thermo Scientific Chemicals Supplier No. L0547718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 776-99-8 | C11H14O3 | 194.23 g/mol

3,4-Dimethoxyphenylacetone was used as starting material in the synthesis of α-methyl-dopa, an important nonproteogenic α-amino acid for pharmaceutical applications. 3,4-Dimethoxyphenylacetone undergoes asymmetric amination catalyzed by Brevibacterium linens IFO 12141 strain to yield corresponding optically active amine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxyphenylacetone was used as starting material in the synthesis of α-methyl-dopa, an important nonproteogenic α-amino acid for pharmaceutical applications. 3,4-Dimethoxyphenylacetone undergoes asymmetric amination catalyzed by Brevibacterium linens IFO 12141 strain to yield corresponding optically active amine

Solubility
Insoluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 776-99-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008772
InChI Key UMYZWICEDUEWIM-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
PubChem CID 69896
IUPAC Name 1-(3,4-dimethoxyphenyl)propan-2-one
SMILES COC1=CC=C(CC(C)=O)C=C1OC

Specifications

Density 1.115
Boiling Point 154°C to 158°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5358
Quantity 50 g
Beilstein 1107410
Solubility Information Insoluble in water.
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxyphenylacetone
EINECSNumber 212-285-0
RTECSNumber UC1795500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only