accessibility menu, dialog, popup

UserName

NaOClO

CAS RN 7758-19-2

IUPAC Name: sodium chlorite
Synonyms: sodium chlorite
Molecular Weight (g/mol): 90.44
Molecular Formula: ClNaO2
InChi Key: UKLNMMHNWFDKNT-UHFFFAOYSA-M
SMILES: [Na+].[O-][Cl]=O
Grade 
  • (2)
  • (1)
  • (3)
  • (4)
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (7)
Percent Purity 
  • (5)
  • (7)
Physical Form 
  • (1)
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (5)
  • (6)
  • (2)
  • (1)
  • (3)
  • (4)
  • (5)
  • (2)
  • (4)
  • (7)
  • (5)
  • (7)
  • (1)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)