missing translation for 'onlineSavingsMsg'
Learn More

Chlorite IC Standard, MilliporeSigma™ Supelco™

Catalog No. 111002672
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002672 100 mL
111002671 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111002672 Supplier Merck Emd Millipore Supplier No. ICS006100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Featured Industries: Environmental

Specifications

CAS 7758-19-2
Concentration 1000 μg/mL in H 2 O
Type Analytical Standard
Quantity 100 mL
Grade Analytical Standard