missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorite, Technical Grade, 80%, LabChem™

Catalog No. LC236201
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC236201 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC236201 Supplier LabChem Supplier No. LC236201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-19-2 , 7647-14-5
Molecular Formula ClNaO2
Molecular Weight (g/mol) 90.438
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
Synonym sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf
PubChem CID 23668197
ChEBI CHEBI:78667
IUPAC Name sodium;chlorite
SMILES [O-]Cl=O.[Na+]

Specifications

Color White
CAS Max % 80, 20
Physical Form Crystals
Quantity 500 g
Linear Formula NaClO2
UN Number UN1496
Solubility Information Soluble in water
Formula Weight 90.44
Percent Purity 80%
Grade Technical
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Carbon monoxide; Carbon dioxide; Hydrogen chloride; Sodium oxide
Chemical Name or Material Sodium Chlorite
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Toxic if swallowed.
Fatal in contact with skin or if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear respiratory protection.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Collect spillage.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature